Assay Details
| Gene Symbol: yae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044480.1 | NP_001037945.1 | 1 - 2 | 272 | 62 | |
| GenBank mRNA | BC157326.1 | - | 1 - 2 | 264 | 62 | |
| BC158959.1 | - | 1 - 2 | 162 | 62 | ||
| CR761419.2 | - | 1 - 2 | 272 | 62 | ||
More Information
Set Membership: |
|

yae1
Genomic Map