Assay Details
| Gene Symbol: racgap1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044482.1 | NP_001037947.1 | 16 - 17 | 1946 | 72 | |
| GenBank mRNA | BC135648.1 | - | 15 - 16 | 1920 | 72 | |
| CR761479.2 | - | 16 - 17 | 1946 | 72 | ||
More Information
Set Membership: |
|

racgap1.2
Genomic Map