Assay Details
| Gene Symbol: racgap1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044482.1 | NP_001037947.1 | 5 - 6 | 719 | 64 | |
| GenBank mRNA | CR761479.2 | - | 5 - 6 | 719 | 64 | |
More Information
Set Membership: |
|

racgap1.2
Genomic Map