Assay Details
| Gene Symbol: nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044493.1 | NP_001037958.1 | 2 - 3 | 324 | 75 | |
| XM_012966681.2 | XP_012822135.1 | 3 - 4 | 752 | 75 | ||
| XM_012966682.2 | XP_012822136.1 | 3 - 4 | 752 | 75 | ||
| XM_012966684.2 | XP_012822138.1 | 3 - 4 | 752 | 75 | ||
| XM_012966685.2 | XP_012822139.1 | 2 - 3 | 513 | 75 | ||
| GenBank mRNA | BC121360.1 | - | 3 - 4 | 723 | 75 | |
| BC170738.1 | - | 2 - 3 | 310 | 75 | ||
| BC170742.1 | - | 2 - 3 | 310 | 75 | ||
| CR760664.2 | - | 2 - 3 | 324 | 75 | ||
More Information
Set Membership: |
|

nptn
Genomic Map