Assay Details
| Gene Symbol: ramac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044495.1 | NP_001037960.1 | 2 - 3 | 385 | 80 | |
| XM_012959161.2 | XP_012814615.1 | 2 - 3 | 688 | 80 | ||
| GenBank mRNA | BC154856.1 | - | 2 - 3 | 321 | 80 | |
| CR760936.2 | - | 2 - 3 | 385 | 80 | ||
More Information
Set Membership: |
|

ramac
Genomic Map