Assay Details
| Gene Symbol: dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044505.1 | NP_001037970.1 | 10 - 11 | 891 | 78 | |
| GenBank mRNA | BC161158.1 | - | 10 - 11 | 927 | 78 | |
| CR761670.2 | - | 10 - 11 | 891 | 78 | ||
More Information
Set Membership: |
|

dbf4
Genomic Map