Assay Details
| Gene Symbol: ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044512.1 | NP_001037977.1 | 1 - 2 | 146 | 60 | |
| GenBank mRNA | CR761896.2 | - | 1 - 2 | 146 | 60 | |
More Information
Set Membership: |
|

ranbp9
Genomic Map