Assay Details
| Gene Symbol: angpt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044515.1 | NP_001037980.1 | 5 - 6 | 496 | 77 | |
| XM_012959169.2 | XP_012814623.1 | 6 - 7 | 590 | 77 | ||
| GenBank mRNA | BC158370.1 | - | 5 - 6 | 512 | 77 | |
| CR761914.2 | - | 5 - 6 | 496 | 77 | ||
More Information
Set Membership: |
|

angpt4
Genomic Map