Assay Details
| Gene Symbol: gnaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044517.1 | NP_001037982.1 | 2 - 3 | 868 | 84 | |
| GenBank mRNA | CR761976.1 | - | 2 - 3 | 868 | 84 | |
More Information
Set Membership: |
|

gnaq
Genomic Map