Assay Details
| Gene Symbol: snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045583.1 | NP_001039048.1 | 3 - 4 | 419 | 102 | |
| XM_012952514.2 | XP_012807968.1 | 3 - 4 | 484 | 102 | ||
| XM_012952515.1 | XP_012807969.1 | 3 - 4 | 471 | 102 | ||
| XM_012952516.1 | XP_012807970.1 | 3 - 4 | 456 | 102 | ||
| XM_012952517.2 | XP_012807971.1 | 3 - 4 | 598 | 102 | ||
| XM_012952518.2 | XP_012807972.1 | 3 - 4 | 595 | 102 | ||
| XM_012952519.2 | XP_012807973.1 | 3 - 4 | 647 | 102 | ||
| XM_012952520.2 | XP_012807974.1 | 3 - 4 | 650 | 102 | ||
| XM_012952521.2 | XP_012807975.1 | 3 - 4 | 402 | 102 | ||
| XM_012952522.2 | XP_012807976.1 | 3 - 4 | 519 | 102 | ||
| XM_012952523.2 | XP_012807977.1 | 3 - 4 | 516 | 102 | ||
| XM_012952524.2 | XP_012807978.1 | 3 - 4 | 632 | 102 | ||
| XM_018097770.1 | XP_017953259.1 | 3 - 4 | 888 | 102 | ||
| XM_018097771.1 | XP_017953260.1 | 3 - 4 | 600 | 102 | ||
| XM_018097772.1 | XP_017953261.1 | 3 - 4 | 885 | 102 | ||
| GenBank mRNA | BC166139.1 | - | 2 - 3 | 369 | 102 | |
| CR848447.2 | - | 3 - 4 | 419 | 102 | ||
More Information
Set Membership: |
|

snx21
Genomic Map