Assay Details
Gene Symbol: dapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045587.1 | NP_001039052.1 | 1 - 2 | 81 | 98 | |
GenBank mRNA | CR848499.2 | - | 1 - 2 | 81 | 98 | |
CR926361.1 | - | 1 - 2 | 79 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
