Assay Details
| Gene Symbol: kitlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045596.1 | NP_001039061.1 | 4 - 5 | 494 | 84 | |
| XM_012959172.2 | XP_012814626.1 | 5 - 6 | 590 | 84 | ||
| XM_012959173.2 | XP_012814627.1 | 5 - 6 | 591 | 84 | ||
| XM_012959174.2 | XP_012814628.1 | 4 - 5 | 551 | 84 | ||
| GenBank mRNA | BC084489.1 | - | 4 - 5 | 499 | 84 | |
| BC167914.1 | - | 4 - 5 | 518 | 84 | ||
| CR855401.2 | - | 4 - 5 | 494 | 84 | ||
More Information
Set Membership: |
|

kitlg
Genomic Map