Assay Details
| Gene Symbol: dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045608.1 | NP_001039073.1 | 4 - 5 | 508 | 65 | |
| XM_012953172.2 | XP_012808626.1 | 4 - 5 | 591 | 65 | ||
| XM_012953173.1 | XP_012808627.1 | 4 - 5 | 571 | 65 | ||
| XM_012953174.2 | XP_012808628.1 | 4 - 5 | 561 | 65 | ||
| GenBank mRNA | BC135708.1 | - | 3 - 4 | 385 | 65 | |
| CR855609.1 | - | 4 - 5 | 508 | 65 | ||
More Information
Set Membership: |
|

dnase2
Genomic Map