Assay Details
| Gene Symbol: sox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045619.2 | NP_001039084.1 | 1 | 1552 | 91 | |
| GenBank mRNA | BC123924.1 | - | 1 | 1552 | 91 | |
| BC135263.1 | - | 1 | 1463 | 91 | ||
| CR926278.2 | - | 1 | 1548 | 91 | ||
More Information
Set Membership: |
|

sox4
Genomic Map