Assay Details
| Gene Symbol: usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045620.1 | NP_001039085.1 | 1 - 2 | 173 | 68 | |
| GenBank mRNA | BC135925.1 | - | 1 - 2 | 174 | 68 | |
| CR926280.2 | - | 1 - 2 | 173 | 68 | ||
More Information
Set Membership: |
|

usp30
Genomic Map