Assay Details
| Gene Symbol: galnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045626.1 | NP_001039091.1 | 5 - 6 | 722 | 55 | |
| GenBank mRNA | BC121347.1 | - | 5 - 6 | 728 | 55 | |
| BC135809.2 | - | 5 - 6 | 695 | 55 | ||
| CR942331.2 | - | 5 - 6 | 722 | 55 | ||
More Information
Set Membership: |
|

galnt16
Genomic Map