Assay Details
| Gene Symbol: gzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045636.1 | NP_001039101.1 | 2 - 3 | 1466 | 67 | |
| XM_012963100.1 | XP_012818554.1 | 3 - 4 | 1499 | 67 | ||
| XM_012963101.2 | XP_012818555.1 | 3 - 4 | 1560 | 67 | ||
| XM_012963102.2 | XP_012818556.1 | 3 - 4 | 1565 | 67 | ||
| GenBank mRNA | BC121586.1 | - | 2 - 3 | 1008 | 67 | |
| BC171267.1 | - | 2 - 3 | 1459 | 67 | ||
| CR942387.2 | - | 2 - 3 | 1466 | 67 | ||
More Information
Set Membership: |
|

gzf1
Genomic Map