Assay Details
| Gene Symbol: smox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045670.1 | NP_001039135.1 | 6 - 7 | 1594 | 57 | |
| GenBank mRNA | CR942507.2 | - | 6 - 7 | 1594 | 57 | |
More Information
Set Membership: |
|

smox
Genomic Map