Assay Details
| Gene Symbol: tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045672.2 | NP_001039137.2 | 5 - 6 | 917 | 70 | |
| XM_012961265.2 | XP_012816719.1 | 5 - 6 | 912 | 70 | ||
| GenBank mRNA | BC157758.1 | - | 5 - 6 | 697 | 70 | |
| CR942511.2 | - | 5 - 6 | 825 | 70 | ||
More Information
Set Membership: |
|

tmem184b
Genomic Map