Assay Details
| Gene Symbol: ier5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045694.1 | NP_001039159.1 | 1 | 849 | 75 | |
| GenBank mRNA | BC135332.1 | - | 1 | 689 | 75 | |
| BC154852.1 | - | 1 | 851 | 75 | ||
| CR942620.2 | - | 1 | 849 | 75 | ||
More Information
Set Membership: |
|

ier5
Genomic Map