Assay Details
| Gene Symbol: mogat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045737.1 | NP_001039202.1 | 5 - 6 | 747 | 84 | |
| XM_012970512.2 | XP_012825966.1 | 5 - 6 | 810 | 84 | ||
| GenBank mRNA | CR942398.2 | - | 5 - 6 | 747 | 84 | |
More Information
Set Membership: |
|

mogat1
Genomic Map