Assay Details
| Gene Symbol: klf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045768.1 | NP_001039233.1 | 1 - 2 | 143 | 62 | |
| GenBank mRNA | CR942695.2 | - | 1 - 2 | 143 | 62 | |
More Information
Set Membership: |
|

klf17
Genomic Map