Assay Details
| Gene Symbol: f2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045769.1 | NP_001039234.1 | 1 - 2 | 112 | 113 | |
| GenBank mRNA | CR942697.2 | - | 1 - 2 | 112 | 113 | |
More Information
Set Membership: |
|

f2rl2
Genomic Map