Assay Details
| Gene Symbol: anp32e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045783.1 | NP_001039248.1 | 1 - 2 | 224 | 73 | |
| XM_012968578.2 | XP_012824032.1 | 1 - 2 | 290 | 73 | ||
| GenBank mRNA | BC160790.1 | - | 1 - 2 | 193 | 73 | |
| CR942787.2 | - | 1 - 2 | 224 | 73 | ||
More Information
Set Membership: |
|

anp32e
Genomic Map