Assay Details
| Gene Symbol: ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078689.1 | NP_001072157.1 | 10 - 11 | 1665 | 60 | |
| GenBank mRNA | BC075466.1 | - | 10 - 11 | 1373 | 60 | |
| BC121566.1 | - | 10 - 11 | 1665 | 60 | ||
| CT030371.1 | - | 10 - 11 | 1387 | 60 | ||
More Information
Set Membership: |
|

ppp2r5a
Genomic Map