Assay Details
| Gene Symbol: rpl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078691.1 | NP_001072159.1 | 3 - 4 | 419 | 76 | |
| GenBank mRNA | BC076693.1 | - | 3 - 4 | 411 | 76 | |
| BC118908.1 | - | 3 - 4 | 419 | 76 | ||
More Information
Set Membership: |
|

rpl7a
Genomic Map