Assay Details
| Gene Symbol: lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078702.1 | NP_001072170.1 | 7 - 8 | 1089 | 68 | |
| XM_012959122.1 | XP_012814576.1 | 7 - 8 | 1016 | 68 | ||
| GenBank mRNA | BC092370.1 | - | 7 - 8 | 1026 | 68 | |
| BC121839.1 | - | 7 - 8 | 1089 | 68 | ||
| CT025390.2 | - | 7 - 8 | 1103 | 68 | ||
More Information
Set Membership: |
|

lmf2
Genomic Map