Assay Details
| Gene Symbol: frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078707.1 | NP_001072175.1 | 5 - 6 | 587 | 80 | |
| XM_012961229.2 | XP_012816683.1 | 6 - 7 | 688 | 80 | ||
| GenBank mRNA | BC098323.1 | - | 5 - 6 | 580 | 80 | |
| BC121203.1 | - | 5 - 6 | 587 | 80 | ||
| BC159081.1 | - | 6 - 7 | 598 | 80 | ||
| CT030398.1 | - | 6 - 7 | 612 | 80 | ||
More Information
Set Membership: |
|

frmd8
Genomic Map