Assay Details
| Gene Symbol: c17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078721.1 | NP_001072189.1 | 2 - 3 | 299 | 68 | |
| XM_012953409.2 | XP_012808863.1 | 2 - 3 | 413 | 68 | ||
| XM_018092148.1 | XP_017947637.1 | 2 - 3 | 409 | 68 | ||
| XM_018092149.1 | XP_017947638.1 | 2 - 3 | 414 | 68 | ||
| GenBank mRNA | BC118723.1 | - | 2 - 3 | 299 | 68 | |
More Information
Set Membership: |
|

c17orf49
Genomic Map