Assay Details
| Gene Symbol: stx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078723.1 | NP_001072191.1 | 9 - 10 | 1237 | 66 | |
| GenBank mRNA | BC118729.1 | - | 9 - 10 | 1237 | 66 | |
| CT030504.1 | - | 9 - 10 | 1261 | 66 | ||
More Information
Set Membership: |
|

stx1a
Genomic Map