Assay Details
| Gene Symbol: fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078734.1 | NP_001072202.1 | 5 - 6 | 498 | 67 | |
| XM_012970524.2 | XP_012825978.1 | 6 - 7 | 639 | 67 | ||
| XM_012970525.2 | XP_012825979.1 | 6 - 7 | 637 | 67 | ||
| GenBank mRNA | BC118754.1 | - | 5 - 6 | 498 | 67 | |
| CT485758.1 | - | 5 - 6 | 448 | 67 | ||
More Information
Set Membership: |
|

fam168b
Genomic Map