Assay Details
| Gene Symbol: tp53i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078755.1 | NP_001072223.1 | 2 - 3 | 306 | 89 | |
| XM_012970532.1 | XP_012825986.1 | 2 - 3 | 183 | 89 | ||
| GenBank mRNA | BC118804.1 | - | 2 - 3 | 306 | 89 | |
More Information
Set Membership: |
|

tp53i3
Genomic Map