Assay Details
| Gene Symbol: rnf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078756.1 | NP_001072224.1 | 6 - 7 | 975 | 63 | |
| XM_012964975.2 | XP_012820429.1 | 7 - 8 | 1139 | 63 | ||
| XM_018094299.1 | XP_017949788.1 | 6 - 7 | 1035 | 63 | ||
| GenBank mRNA | BC118805.1 | - | 6 - 7 | 975 | 63 | |
More Information
Set Membership: |
|

rnf34
Genomic Map