Assay Details
| Gene Symbol: rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078757.1 | NP_001072225.1 | 4 - 5 | 1154 | 89 | |
| GenBank mRNA | BC118806.1 | - | 4 - 5 | 1154 | 89 | |
| BC135390.1 | - | 5 - 6 | 533 | 89 | ||
More Information
Set Membership: |
|

rabac1
Genomic Map