Assay Details
| Gene Symbol: vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078762.1 | NP_001072230.1 | 4 - 5 | 565 | 66 | |
| GenBank mRNA | BC118814.1 | - | 4 - 5 | 565 | 66 | |
More Information
Set Membership: |
|

vti1a
Genomic Map