Assay Details
| Gene Symbol: igsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078772.2 | NP_001072240.2 | 589 | 57 | ||
| GenBank mRNA | BC118837.1 | - | 589 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

igsf21| Gene Symbol: igsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078772.2 | NP_001072240.2 | 589 | 57 | ||
| GenBank mRNA | BC118837.1 | - | 589 | 57 | ||
Set Membership: |
|