Assay Details
| Gene Symbol: ttc9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078791.2 | NP_001072259.2 | 2 - 3 | 617 | 91 | |
| GenBank mRNA | BC121245.1 | - | 2 - 3 | 605 | 91 | |
| CU075823.1 | - | 2 - 3 | 615 | 91 | ||
More Information
Set Membership: |
|

ttc9b
Genomic Map