Assay Details
| Gene Symbol: wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078795.1 | NP_001072263.1 | 7 - 8 | 1122 | 64 | |
| XM_018091335.1 | XP_017946824.1 | 7 - 8 | 1212 | 64 | ||
| XM_018091336.1 | XP_017946825.1 | 8 - 9 | 1349 | 64 | ||
| GenBank mRNA | BC121252.1 | - | 7 - 8 | 1122 | 64 | |
More Information
Set Membership: |
|

wasf3
Genomic Map