Assay Details
| Gene Symbol: nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078798.1 | NP_001072266.1 | 11 - 12 | 1514 | 62 | |
| XM_018094693.1 | XP_017950182.1 | 10 - 11 | 1416 | 62 | ||
| XM_018094694.1 | XP_017950183.1 | 10 - 11 | 1448 | 62 | ||
| GenBank mRNA | BC121262.1 | - | 11 - 12 | 1514 | 62 | |
| CT027917.1 | - | 11 - 12 | 1546 | 62 | ||
More Information
Set Membership: |
|

nmt1
Genomic Map