Assay Details
| Gene Symbol: taf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078817.1 | NP_001072285.1 | 2 - 3 | 1190 | 62 | |
| XM_012952548.2 | XP_012808002.1 | 2 - 3 | 1759 | 62 | ||
| GenBank mRNA | BC121308.1 | - | 2 - 3 | 1190 | 62 | |
More Information
Set Membership: |
|

taf4
Genomic Map