Assay Details
| Gene Symbol: cntd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078818.1 | NP_001072286.1 | 2 - 3 | 100 | 105 | |
| GenBank mRNA | BC121310.1 | - | 2 - 3 | 100 | 105 | |
| CU024973.1 | - | 99 | 105 | |||
More Information
Set Membership: |
|

cntd2
Genomic Map