Assay Details
Gene Symbol: cntd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078818.1 | NP_001072286.1 | 6 - 7 | 601 | 92 | |
GenBank mRNA | BC121310.1 | - | 6 - 7 | 601 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
