Assay Details
| Gene Symbol: smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078823.1 | NP_001072291.1 | 2 - 3 | 1733 | 96 | |
| GenBank mRNA | BC121322.1 | - | 2 - 3 | 1733 | 96 | |
| CT025291.2 | - | 2 - 3 | 1735 | 96 | ||
More Information
Set Membership: |
|

smg8
Genomic Map