Assay Details
| Gene Symbol: pdzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078826.1 | NP_001072294.1 | 910 | 75 | ||
| GenBank mRNA | BC121328.1 | - | 910 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdzd8| Gene Symbol: pdzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078826.1 | NP_001072294.1 | 910 | 75 | ||
| GenBank mRNA | BC121328.1 | - | 910 | 75 | ||
Set Membership: |
|