Assay Details
| Gene Symbol: nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078835.1 | NP_001072303.1 | 5 - 6 | 1482 | 82 | |
| XM_012957242.2 | XP_012812696.1 | 5 - 6 | 1540 | 82 | ||
| XM_012957243.2 | XP_012812697.1 | 5 - 6 | 1435 | 82 | ||
| XM_012957244.2 | XP_012812698.1 | 5 - 6 | 1667 | 82 | ||
| GenBank mRNA | BC121356.1 | - | 5 - 6 | 1482 | 82 | |
More Information
Set Membership: |
|

nr4a1
Genomic Map