Assay Details
| Gene Symbol: gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078848.1 | NP_001072316.1 | 8 - 9 | 1913 | 63 | |
| GenBank mRNA | BC121374.1 | - | 8 - 9 | 1913 | 63 | |
More Information
Set Membership: |
|

gpc1
Genomic Map