Assay Details
| Gene Symbol: bicdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078860.1 | NP_001072328.1 | 9 - 10 | 1816 | 89 | |
| GenBank mRNA | BC121401.1 | - | 9 - 10 | 1816 | 89 | |
More Information
Set Membership: |
|

bicdl1
Genomic Map