Assay Details
| Gene Symbol: mapk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078864.1 | NP_001072332.1 | 5 - 6 | 2761 | 64 | |
| XM_012970540.2 | XP_012825994.1 | 5 - 6 | 2754 | 64 | ||
| XM_012970541.2 | XP_012825995.1 | 5 - 6 | 2591 | 64 | ||
| XM_012970542.2 | XP_012825996.1 | 5 - 6 | 2670 | 64 | ||
| GenBank mRNA | BC121410.1 | - | 5 - 6 | 2761 | 64 | |
More Information
Set Membership: |
|

mapk7
Genomic Map