Assay Details
| Gene Symbol: klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078869.1 | NP_001072337.1 | 4 - 5 | 1465 | 64 | |
| GenBank mRNA | BC121419.1 | - | 4 - 5 | 1465 | 64 | |
| BC160560.1 | - | 4 - 5 | 1376 | 64 | ||
More Information
Set Membership: |
|

klhl24
Genomic Map