Assay Details
| Gene Symbol: asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078873.1 | NP_001072341.1 | 178 | 61 | ||
| XM_012965665.2 | XP_012821119.1 | 1 - 2 | 355 | 61 | ||
| GenBank mRNA | BC121423.1 | - | 178 | 61 | ||
More Information
Set Membership: |
|

asna1
Genomic Map